missing translation for 'onlineSavingsMsg'
Learn More

Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 75 to 125 μm, Purolite™

Product Code. 17029463
Click to view available options
Chemical Name or Material:
Diethylamino Methacrylate Macroporous Weak Anion Exchanger
Epoxy Methacrylate Macroporous Affinity Resin
Iminodiacetic Acid Methacrylate Affinity Resin
Quaternary Ammonium Methacrylate Macroporous Strong Anion Exchanger
Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger
Physical Form:
Spherical Beads
Wet
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 17029463

Brand: Chromalite LS0405294

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.

Hydrophilic, macroporous resins based on a methacrylic backbone.

  • Ion exchange chromatography
  • Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
  • Suitable for large-scale applications
  • Excellent pressure-flow properties,designed for high performance, stability and reliability
  • Purification of peptides obtained by fermentation
  • High mechanical stability
  • Uniform particle size
  • Stable to CIP (up to 1M NaOH)
  • Negligible swelling
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free

Specifications

Chemical Name or Material Iminodiacetic Acid Methacrylate Affinity Resin
Physical Form Spherical Beads
Particle Size 75 to 125 μm
Pore Size 1000 Å
Color White to Off-white
Quantity 500 mL
Product Line Chromalite™ MIDA/M
pH Range 1 to 14
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.