missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 100 to 300 μm, Purolite™
Description
Hydrophilic, macroporous resins based on a methacrylic backbone.
- Ion exchange chromatography
- Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
- Suitable for large-scale applications
- Excellent pressure-flow properties,designed for high performance, stability and reliability
- Purification of peptides obtained by fermentation
- High mechanical stability
- Uniform particle size
- Stable to CIP (up to 1M NaOH)
- Negligible swelling
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
| Chemical Name or Material | Epoxy Methacrylate Macroporous Affinity Resin |
| Physical Form | Wet |
| Particle Size | 100 to 300 μm |
| Pore Size | 1000 Å |
| Quantity | 100 mL |
| Product Line | Chromalite™ MEP/C |
| Functionality/Interaction Type | Epoxy |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?