missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Chromalite™ HPLC Column for RP and IEX Chromatography
Click to view available options
Packing Material:
Chromalite™ 10AD2
Chromalite™ 10AD2Q
Chromalite™ 10AD2S
Chromalite™ 5AD2
Dimensions:
4.6 mm x 150 mm
4.6 mm x 250 mm
Unit Size:
Each
Description
Analytical Reverse Phase chromatography (RP-HPLC) and ion-exchange chromatography (IEX-HPLC) resins for separation of proteins, peptides and oligonucleotides, and ideal for biomolecule purification and polishing.
- Uniform particle sizes
- Exceptional batch-to-batch consistency
- Simple scale-up from laboratory analyses through to full-scale plant operation
- Highly hydrophobic
- High chemical stability
- More robust than silica materials
- Pressure stability up to 50 bar
- Efficient regeneration
- Pre-packed in stainless steel HPLC columns
- Compliant with FDA regulation 21 CFR 173.65
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
| Particle Size | 4 to 6 μm |
| pH | 1 to 14 pH |
| Pore Size | 200 to 300 Å |
| Surface Area | >500 m2/g |
| Content And Storage | 5 years from date of manufacture; Storage (at 2°C to 20°C) |
| Dimensions | 4.6 mm x 150 mm |
| Packing Material | Chromalite™ 5AD2 |
| Material | Stainless Steel |
| Type | Macroporous Polystyrene/Divinylbenzene Adsorbent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction