missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chromacol 9mm Screw Cas
Click to view available options
Color:
Black
Pre-slit:
No
Material (Liner):
Prefitted Silicone/PTFE
Prefitted Type 8 Rubber/PTFE
Unit Size:
Pack of 500
Description
Thermo Scientific™ Chromacol 9mm Screw Caps are offered with a selection of liner materials and colors.
Specifications
Specifications
| Description | 9mm Screw Cap with Flange for Japanese Instruments Prefitted Type 8 Rubber/PTFE Liner |
| Material (Liner) | Prefitted Type 8 Rubber/PTFE |
| Color | Black |
| Material | Prefitted Type 8 Rubber |
| Pre-slit | No |
| Product Line | Chromacol |
| Closure Size | 9 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction