missing translation for 'onlineSavingsMsg'
Learn More

ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™

ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines.

Brand:  Thermo Scientific T111102500

Additional Details : Weight : 2.50000kg Transport : UN number : 1992 Chem class : 3 Pack group : II

 View more versions of this product

Product Code. 15803197

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
  • Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
  • Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
  • A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
  • Offered in 1L borosilicate bottles and 2.5L amber glass bottles
Specifications

Specifications

Calcium (Ca): ≤ 20 ppb Sodium (Na): ≤ 50 ppb
75-05-8; 67-56-1; 67-63-0; 7732-18-5
Colorless
LC-MS
Amber glass bottle
2.5 L
25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA)
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only