Learn More
Macherey Nagel CHROMABOND columns Flash RS 40filled with: ALOX N filling: 80 g Material: PP,
Brand:  Macherey Nagel 732881
CHROMABOND columns Flash RS 40filled with: ALOX N filling: 80 g Material: PP,
Product Code. 12788362
  • £480.00 / Pack of 1
Only null left
Estimated Shipment: 26-05-2026