Learn More
Central Surgical Company™ Chroma Gesellshaft™ Stainless Steel Needle
Click to view available options
Needle Gauge:
14 G
16 G
19 G
Unit Size:
Pack of 10
Specifications
Specifications
| Material | Stainless Steel |
| Needle Gauge | 19 G |
| Product Type | Needle |
| Length (Metric) Needle | 101 mm |
| Needle Point Style | Long |
| For Use With (Equipment) | Hypodermic Syringe Long Luer Lock Fitting |