missing translation for 'onlineSavingsMsg'
Learn More

Azpack Tape printed STERILE, on roll, text repeated along tape, 12mm width, 66m roll

Brand:  Azpack LAB-785-E

Tape printed STERILE, on roll, text repeated along tape, 12mm width, 66m roll

Code : 02

Additional Details : Weight : 0.04888kg

Product Code. 10234534

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.