missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103556
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CHRNG.
The mammalian muscle-type acetylcholine receptor is a transmembrane pentameric glycoprotein with two alpha subunits, one beta, one delta, and one epsilon (in adult skeletal muscle) or gamma (in fetal and denervated muscle) subunit. This gene, which encodes the gamma subunit, is expressed prior to the thirty-third week of gestation in humans. The gamma subunit of the acetylcholine receptor plays a role in neuromuscular organogenesis and ligand binding and disruption of gamma subunit expression prevents the correct localization of the receptor in cell membranes. Mutations in this gene cause Escobar syndrome and a lethal form of multiple pterygium syndrome. Muscle-type acetylcholine receptor is the major antigen in the autoimmune disease myasthenia gravis.
Specifications
| CHRNG | |
| Polyclonal | |
| Unconjugated | |
| CHRNG | |
| acetylcholine receptor; Acetylcholine receptor subunit gamma; acetylcholine receptor, muscle, gamma subunit; acetylcholine receptor, nicotinic, gamma (muscle); AChR gamma subunit; Achr-3; ACHRG; Acrg; cholinergic receptor; cholinergic receptor nicotinic gamma subunit; cholinergic receptor, nicotinic gamma; cholinergic receptor, nicotinic, gamma (muscle); cholinergic receptor, nicotinic, gamma polypeptide; Chrng; chrng variant N1; chrng variant N2; nicotinic acetylcholine receptor gamma subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11449, 1146, 25753 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04760, P07510, P18916 | |
| CHRNG | |
| A synthesized peptide derived from human CHRNG(Accession P07510), corresponding to amino acid residues A20-E70. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction