missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRND Antibody (1H1F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61733-0.025ml
This item is not returnable.
View return policy
Description
CHRND Monoclonal specifically detects CHRND in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| CHRND | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| acetylcholine receptor subunit delta, ACHRDSCCMS, cholinergic receptor, nicotinic, delta, cholinergic receptor, nicotinic, delta polypeptide, CMS2A, FCCMS | |
| Mouse | |
| 58.8 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 1H1F9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CHRND | |
| Purified recombinant fragment of human CHRND (AA: extra 22-245) expressed in E. Coli. | |
| Protein G purified | |
| GPCR, Hypoxia | |
| 1144 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction