missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNB4 Monoclonal Antibody (1H11F2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531695
This item is not returnable.
View return policy
Description
MA5-31695 has been tested in indirect ELISA.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications
| CHRNB4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P30926 | |
| CHRNB4 | |
| Purified recombinant fragment of human CHRNB4 (AA: extra 22-236) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 1H11F2 | |
| Unconjugated | |
| CHRNB4 | |
| acetylcholine receptor; Acetylcholine receptor beta 4; acetylcholine receptor, nicotinic, beta 4 (neuronal); Acrb4; Acrb-4; cholinergic receptor; cholinergic receptor nicotinic beta 4 subunit; cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Chrnb4; nAChRB4; N-alpha 2; neuronal acetylcholine receptor non-alpha-2 chain; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit; nicotinic acetylcholine receptor beta4 subunit | |
| Mouse | |
| Protein G | |
| RUO | |
| 1143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction