missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117458
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
The nicotinic acetylcholine receptors are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are stated that the proposed structure for each subunit is a conserved N-terminal extracellular domain followed by 3 conserved transmembrane domains, a variable cytoplasmic loop, a fourth conserved transmembrane domain, and a short C-terminal extracellular region.
Specifications
| CHRNB3 | |
| Polyclonal | |
| Unconjugated | |
| Chrnb3 | |
| 5730417K16Rik; acetylcholine receptor; acetylcholine receptor, neuronal nicotinic, beta-3 subunit; acetylcholine receptor, nicotinic, beta 3 (neuronal); Acrb3; cholinergic receptor; cholinergic receptor nicotinic beta 3 subunit; cholinergic receptor, nicotinic beta 3; cholinergic receptor, nicotinic, beta 3 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 3; Chrnb3; I79_003985; Neuronal acetylcholine receptor subunit beta-3; neuronal nicotinic | |
| Rabbit | |
| Protein G | |
| RUO | |
| 108043, 1142 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q05901, Q8BMN3 | |
| Chrnb3 | |
| Recombinant Human Neuronal acetylcholine receptor subunit beta-3 protein (67-218aa). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction