missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11553-1-AP-150UL
This item is not returnable.
View return policy
Description
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.Specifications
| CHRNB1 | |
| Polyclonal | |
| Unconjugated | |
| CHRNB1 | |
| ACHRB, CHRNB, CHRNB1, CMS1D, CMS2A, SCCMS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1140, 11443, 24261 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09690, P11230, P25109 | |
| CHRNB1 | |
| CHRNB1 Fusion Protein Ag2108 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction