missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101398
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CHRNB1.
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
Specifications
| CHRNB1 | |
| Polyclonal | |
| Unconjugated | |
| Chrnb1 | |
| acetylcholine receptor; acetylcholine receptor beta; acetylcholine receptor subunit beta; acetylcholine receptor, nicotinic, beta 1 (muscle); AChR beta; Achr-2; ACHRB; Acrb; cholinergic receptor; cholinergic receptor nicotinic beta 1 subunit; cholinergic receptor nicotinic beta polypeptide 1; cholinergic receptor, nicotinic beta 1; cholinergic receptor, nicotinic, beta 1 (muscle); cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); CHRNB; Chrnb 1; CHRNB1; CMS1D; CMS2A; CMS2C; nAChRB1; RNACRB1; SCCMS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1140, 11443, 24261 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09690, P11230, P25109 | |
| Chrnb1 | |
| A synthesized peptide derived from human CHRNB1(Accession P11230), corresponding to amino acid residues L63-V113. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction