Learn More
CHRNA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26025-1-AP-150UL
 View more versions of this product
Product Code. 16834555
  • £349.00 / 150µL
Description
CHRNA9 is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor superfamily. CHRNA9 is a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. It is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea.This gene is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor gene superfamily. It encodes a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. This protein is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea. The protein is additionally expressed in keratinocytes, the pituitary gland, B-cells and T-cells.
Specifications
Specifications
CHRNA9
Polyclonal
Unconjugated
CHRNA9
CHRNA9, HSA243342, NACHR alpha 9, NACHRA9
Rabbit
Antigen Affinity Chromatography
RUO
231252, 55584
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UGM1
CHRNA9
CHRNA9 Fusion Protein Ag23272
150 μL
Primary
Human, Mouse