missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRNA4 Recombinant Rabbit Monoclonal Antibody (K01_2N54)

Product Code. 30829591
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829591 50 μL
30827077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829591 Supplier Invitrogen™ Supplier No. MA565599
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CHRNA4 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA4
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N54
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene CHRNA4
Gene Accession No. O70174, P09483, P43681
Gene Alias a4 nicotinic receptor; acetylcholine receptor; acetylcholine receptor alpha 4 neural; acetylcholine receptor, neuronal, nicotinic, alpha-4 subunit; Acra4; Acra-4; alpha4 nAChR; alpha4-nAChR; BFNC; cholinergic receptor; cholinergic receptor nicotinic alpha 4 subunit; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha 4 subunit; cholinergic receptor, nicotinic, alpha polypeptide 4; CHRNA 4; Chrna4; EBN; EBN1; ENFL1; NACHR; nAChR alpha 4; NACHRA4; NACRA4; NARAC; neuronal acetylcholine receptor subunit alpha-4; Neuronal nicotinic acetylcholine receptor alpha 4 subunit; neuronal nicotinic acetylcholine receptor alpha-4 subunit
Gene Symbols CHRNA4
Host Species Rabbit
Immunogen A synthetic peptide of human Nicotinic Acetylcholine Receptor alpha 4/CHRNA4.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100760001, 1137, 11438, 25590
Target Species Human, Mouse, Rat, Hamster
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.