missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNA4 Recombinant Rabbit Monoclonal Antibody (23GB5085), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552784
This item is not returnable.
View return policy
Description
This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described.
Specifications
| CHRNA4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CHRNA4 | |
| a4 nicotinic receptor; acetylcholine receptor; acetylcholine receptor alpha 4 neural; acetylcholine receptor, neuronal, nicotinic, alpha-4 subunit; Acra4; Acra-4; alpha4 nAChR; alpha4-nAChR; BFNC; cholinergic receptor; cholinergic receptor nicotinic alpha 4 subunit; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha 4 subunit; cholinergic receptor, nicotinic, alpha polypeptide 4; CHRNA 4; Chrna4; EBN; EBN1; ENFL1; NACHR; nAChR alpha 4; NACHRA4; NACRA4; NARAC; neuronal acetylcholine receptor subunit alpha-4; Neuronal nicotinic acetylcholine receptor alpha 4 subunit; neuronal nicotinic acetylcholine receptor alpha-4 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1137 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5085 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43681 | |
| CHRNA4 | |
| A synthesized peptide derived from human ACHA4 (540-600AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction