missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577502
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described.
Specifications
| CHRNA4 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| CHRNA4 | |
| nAChR alpha 4, CHRNA4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1137, 11438, 25590 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O70174, P09483, P43681 | |
| Chrna4 | |
| Peptide (C)DLVNMHSRVDQLD, corresponding to amino acid residues 190-202 of human nAChRalpha4 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction