Learn More
CHRNA4 293T Cell Overexpression Lysate (Denatured), Abnova
Description
This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described.
- Transfected Cell Line: 293T
- Plasmid: pCMV-CHRNA4 full-length
- Host: Human
- Storage Buffer: 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | CHRNA4 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC096292.1 |
| Molecular Weight (g/mol) | 49.72 |
| pH Range | 6.8 |
| Plasmid | pCMV-CHRNA4 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-CHRNA4 antibody (H00001137-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.