missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10333-1-AP-150UL
This item is not returnable.
View return policy
Description
CHRNA3 locus encodes a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.Specifications
| CHRNA3 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA3 | |
| CHRNA3, LNCR2, NACHRA3, PAOD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110834, 1136 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P32297, Q8R4G9 | |
| Chrna3 | |
| CHRNA3 Fusion Protein Ag0272 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction