Learn More
CHRNA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556144
Description
Immunogen sequence: RTPTTHTMPS WVKTVFLNLL PRVMFMTRPT SNEGNAQKPR PLYGAELSNL NCFSRAESKG CKEGYPCQDG MCGYCHHRRI KISNFSANLT RSSSSESVDA VLSLSALSPE IKEAIQSVKY IAENMKAQNE AKEIQDDWKY VAM Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
CHRNA3 locus encodes a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.
Specifications
| CHRNA3 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA3 | |
| LNCR2, NACHRA3, PAOD2, acetylcholine receptor, nicotinic, alpha 3 (neuronal), cholinergic receptor, nicotinic, alpha polypeptide 3, neuronal acetylcholine receptor subunit alpha-3, neuronal nicotinic acetylcholine receptor, alpha3 subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1136 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P32297 | |
| CHRNA3 | |
| Recombit protein corresponding to Human CHRNA3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only