missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115016
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CHRM4.
Muscarinic acetylcholine receptor M4 is an Acetylcholine Receptor. Unlike muscarinic receptor M2, the M4 receptor is not involved in smooth muscle contraction. The clinical relevance of this receptor is unknown. However, mouse studies have linked it to adenylyl cyclase inhibition. Muscarinic acetylcholine receptor M4 has been reported to be expressed in many regions of the rat brain, particularly the striatum and hippocampus. ESTs have been isolated from human breast and eye libraries.
Specifications
| CHRM4 | |
| Polyclonal | |
| Unconjugated | |
| CHRM4 | |
| acetylcholine receptor; acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor; cholinergic receptor muscarinic 4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Chrm-4; HM4; M4; M4R; Mm4 mAChR; muscarinic acetylcholine receptor 4; muscarinic acetylcholine receptor M4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1132, 12672, 25111 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P08173, P08485, P32211 | |
| CHRM4 | |
| A synthesized peptide derived from human CHRM4(Accession P08173), corresponding to amino acid residues A243-S293. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction