Learn More
CHRM2 Mouse anti-Human, Mouse, Clone: 1424CT461.78.60, Abnova™
Mouse monoclonal antibody raised against human CHRM2.
Brand: Abnova MAB12363.400uL
Description
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine to these receptors and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 2 is involved in mediation of bradycardia and a decrease in cardiac contractility. Multiple alternatively spliced transcript variants have been described for this gene. [provided by RefSeq]
Specifications
| CHRM2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1:25) Immunohistochemistry (1:25) Immunofluorescence (1:25) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| FLJ43243/HM2/MGC120006/MGC120007 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 1129 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1424CT461.78.60 | |
| Mouse monoclonal antibody raised against human CHRM2. | |
| In PBS (0.09% sodium azide) | |
| CHRM2 | |
| Recombinant protein corresponding to human CHRM2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.