missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588481
This item is not returnable.
View return policy
Description
Immunogen sequence: WELGYWLCYV NSTINPMCYA LCNKAFRDTF RLLLLCRWDK RRWRKIPKRP GSVHRTPSRQ C; Positive Samples: U-87MG, HepG2; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 1 is involved in mediation of vagally-induced bronchoconstriction and in the acid secretion of the gastrointestinal tract. The gene encoding this receptor is localized to 11q13.
Specifications
| CHRM1 | |
| Polyclonal | |
| Unconjugated | |
| CHRM1 | |
| acetylcholine receptor; acetylcholine receptor, muscarinic 1; AChR M1; cholinergic receptor; cholinergic receptor muscarinic 1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; cholinergic receptor, muscarinic 1, CNS; CHRM1; Chrm-1; HM1; M1; M1 mAChR; m1 muscarinic acetylcholine receptor protein; M1R; MGC30125; muscarinic acetylcholine receptor M1; muscarinic receptor M1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1128, 12669, 25229 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P08482, P11229, P12657 | |
| CHRM1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-460 of human CHRM1 (NP_0007292). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction