missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5120489

Product Code. 17791884

  • £429.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse. Immunogen sequence: PPAVSPNITV LAPGKGPWQ.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 1 is involved in mediation of vagally-induced bronchoconstriction and in the acid secretion of the gastrointestinal tract. The gene encoding this receptor is localized to 11q13.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHRM1
Polyclonal
Unconjugated
CHRM1
acetylcholine receptor; acetylcholine receptor, muscarinic 1; AChR M1; cholinergic receptor; cholinergic receptor muscarinic 1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; cholinergic receptor, muscarinic 1, CNS; CHRM1; Chrm-1; HM1; M1; M1 mAChR; m1 muscarinic acetylcholine receptor protein; M1R; MGC30125; muscarinic acetylcholine receptor M1; muscarinic receptor M1
Rabbit
Affinity chromatography
RUO
1128, 12669, 25229
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
2.23 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P08482, P11229, P12657
CHRM1
Synthetic peptide
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.