missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRFAM7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35255-100ul
This item is not returnable.
View return policy
Description
CHRFAM7A Polyclonal antibody specifically detects CHRFAM7A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CHRFAM7A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CHRNA7, CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (familywith sequence similarity 7A, exons A-E) fusion, CHRNA7 (cholinergic receptor, nicotinic, alpha polypeptide 7, exons 5-10) andFAM7A (family with sequence similarity 7A, exons A-E) fusion, CHRNA7-DR1alpha 7 neuronal nicotinic acetylcholine receptor-FAM7A hybrid, CHRNA7-FAM7A fusion protein, D-10alpha-7 nicotinic cholinergic receptor subunit, MGC120482, MGC120483 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CHRFAM7A (NP_647536.1).,, Sequence:, MQKYCIYQHFQFQLLIQHLWIAANCDIADERFDATFHTNVLVNSSGHCQYLPPGIFKSSCYIDVRWFPFDVQHCKLKFGSWSYGGWSLDLQMQEADISGYIPNGEWDLVGIPGKRSERFYECCKEPYPDVTFTVTMRRRTLYYGLNLLIP | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 89832 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction