missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHREBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56612-25ul
This item is not returnable.
View return policy
Description
CHREBP Polyclonal specifically detects CHREBP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CHREBP | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14°Carbohydrate response element binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MLXIPL | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GSERRLSGDLSSMPGPGTLSVRVSPPQPILSRGRPDSNKTENRRITHISAE | |
| 25 μL | |
| Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators | |
| 51085 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction