missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHREBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92977-0.1ml
This item is not returnable.
View return policy
Description
CHREBP Polyclonal antibody specifically detects CHREBP in Human, Mouse samples. It is validated for Western Blot
Specifications
| CHREBP | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-175 of human MLXIPL (NP_116569.1). VAPSPDSDSDTDSEDPSLRRSAGGLLRSQVIHSGHFMVSSPHSDSLPRRRDQEGSVGPSDFGPRSIDPTLTRLFECLSLAYSGKLVSPKWKNFKGLKLLCRDKIRLNNAIWRAWYIQYVKRRKSPVCGFVTPLQGPEADAHRKPEAVVLEGNYWKRRIE | |
| 0.1 mL | |
| Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators | |
| 51085 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction