missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRDL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119023
This item is not returnable.
View return policy
Description
CHRDL1 Polyclonal Antibody for Western Blot, IHC (P)
CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| CHRDL1 | |
| Polyclonal | |
| Unconjugated | |
| CHRDL1 | |
| CHL; CHL1; chordin like 1; chordin-like 1; chordin-like protein 1; Chrdl1; dA141H5.1; Kjr; Kohjirin; MGC1; MGCN; neuralin 1; neuralin-1; neurogenesin 1; Neurogenesin-1; Ng1; Nrln1; Ventroptin; ventroptin-alpha; ventroptin-beta; VOPT | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 363455, 83453, 91851 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q76LD0, Q920C1, Q9BU40 | |
| CHRDL1 | |
| Recombinant protein Chordin Like Protein 1. The antigen corresponds to amino acid range 272-447 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction