missing translation for 'onlineSavingsMsg'
Learn More

CHR-6494, MedChemExpress

Product Code. 16416676
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416676

Brand: Medchem Express HY15217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CHR-6494 is a potent inhibitor of haspin, with an IC50 of 2 nM. CHR-6494 inhibits histone H3T3 phosphorylation. CHR-6494 can be used in the research of cancer.

CHR-6494 is a potent inhibitor of haspin, with an IC50 of 2 nM. CHR-6494 inhibits histone H3T3 phosphorylation. CHR-6494 can be used in the research of cancer.

Specifications

Chemical Name or Material CHR-6494
CAS 1333377-65-3
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C16H16N6
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (171.03 mM; Need ultrasonic)
SMILES CCCNC(C=C1)=NN2C1=NC=C2C3=CC(C=NN4)=C4C=C3
Molecular Weight (g/mol) 292.34
Formula Weight 292.34
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.