missing translation for 'onlineSavingsMsg'
Learn More

CHR-6494, MedChemExpress

Product Code. 16466368
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466368 10 mg 10mg
16458957 5 mg 5mg
16416676 100 mg 100mg
16412487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466368 Supplier Medchem Express Supplier No. HY15217
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CHR-6494 is a potent inhibitor of haspin, with an IC50 of 2 nM. CHR-6494 inhibits histone H3T3 phosphorylation. CHR-6494 can be used in the research of cancer.

CHR-6494 is a potent inhibitor of haspin, with an IC50 of 2 nM. CHR-6494 inhibits histone H3T3 phosphorylation. CHR-6494 can be used in the research of cancer.

Specifications

Chemical Name or Material CHR-6494
CAS 1333377-65-3
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C16H16N6
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (171.03 mM; Need ultrasonic)
SMILES CCCNC(C=C1)=NN2C1=NC=C2C3=CC(C=NN4)=C4C=C3
Molecular Weight (g/mol) 292.34
Formula Weight 292.34
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.