missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHPT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92299-0.02ml
This item is not returnable.
View return policy
Description
CHPT1 Polyclonal antibody specifically detects CHPT1 in Human samples. It is validated for Western Blot
Specifications
| CHPT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| AAPT1-like protein, choline phosphotransferase 1, cholinephosphotransferase 1 alpha, CPT, CPT1cholinephosphotransferase 1, Diacylglycerol cholinephosphotransferase 1, EC 2.7.8, EC 2.7.8.2, hCPT1, phosphatidylcholine synthesizing enzyme | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CHPT1 (NP_064629.2). MAAGAGAGSAPRWLRALSEPLSAAQLRRLEEHRYSAAGVSLLEPPLQLYWTWLLQWIPLWMAPNSITLLGLAVNVVTTLVLISYCPTATEEAPYWTYLLC | |
| 0.02 mL | |
| Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 56994 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction