missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHPF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557164
This item is not returnable.
View return policy
Description
Immunogen sequence: GCTGDHEGVH YSHLELSPGE PVQEGDPHFR SALTAHPVRD PVHMYQLHKA FARAELERTY QEIQELQWEI QNTSHLAVDG DRAAAWPVGI PAPS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.
Chondroitin sulfate synthases (CHSYs) synthesize chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to various of core protein families and regulate many biologic processes, including extracellular matrix deposition, cell proliferation and recognition, and morphogenesis. The CHSY family includes CHSY1, CHSY2 and CHSY3. CHSY1 and CHSY3 display both glucuronyltransferase and N-acetylgalactosaminyltransferase activities, while CHSY2 is required for chondroitin polymerizing activity. CHSY2 localizes to the Golgi apparatus and is expressed ubiquitously, with highest expression observed in pancreas, ovary, brain, heart, skeletal muscle, colon, kidney, liver, stomach, small intestine and placenta.
Specifications
| CHPF | |
| Polyclonal | |
| Unconjugated | |
| CHPF | |
| 1700028N03Rik; AI414328; Chondroitin glucuronyltransferase 2; chondroitin polymerizing factor; chondroitin sulfate synthase 2; Chondroitin-polymerizing factor; CHPF; CHSY2; CSS2; D1Bwg1363e; glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II; N-acetylgalactosaminyltransferase 2; UNQ651/PRO1281 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZ52 | |
| CHPF | |
| Recombinant protein corresponding to Human CHPF. Recombinant protein control fragment (Product #RP-96062). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction