missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101475
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CHP2.
CHP2 (Calcineurin B homologous protein 2), also known as Hepatocellular carcinoma-associated antigen 520, is a 196 amino acid protein that contains four EF-hand domains and plays a potential role in transmembrane Na+/H+ exchange. By binding to and activating NHE-1, CHP2 increases the pH and works to protect cells from serum deprivation-induced death. Though not typically detected in normal tissues, CHP2 is highly expressed in malignantly transformed cells and is therefore considered to be a tumor-associated antigen. Ectopic expression of CHP2 promotes proliferation of HEK293 cells and knockdown of CHP2 mRNA in HepG2 cells inhibits cell proliferation. Like calcineurin B, CHP2 can bind to and stimulate phosphatase activity of calcineurin A and activate the calcineurin/NFAT signaling pathway.
Specifications
| CHP2 | |
| Polyclonal | |
| Unconjugated | |
| CHP2 | |
| 2010110P09Rik; calcineurin B homologous protein 2; calcineurin like EF-hand protein 2; calcineurin-like EF hand protein 2; calcineurin-like EF-hand protein 2; Cbhp2; CHP2; chp-2; HCA520; hepatocellular carcinoma antigen gene 520; hepatocellular carcinoma-associated antigen 520; Hepatocellular carcinoma-associated antigen 520 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 308965, 63928, 70261 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43745, Q810D1, Q9D869 | |
| CHP2 | |
| A synthesized peptide derived from human CHP2(Accession O43745), corresponding to amino acid residues P103-L153. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction