missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chp2 Antibody (CPTC-CHP2-1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08694PE
This item is not returnable.
View return policy
Description
Chp2 Monoclonal antibody specifically detects Chp2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Chp2 | |
| Monoclonal | |
| PE | |
| calcineurin B homologous protein 2, HCA520, hepatocellular carcinoma antigen gene 520, Hepatocellular carcinoma-associated antigen 520 | |
| Recombinant human full-length Chp2 protein (Uniprot: O43745) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CPTC-CHP2-1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 63928 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction