missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA529876

Product Code. 13464217

  • £481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HeLa, HepG2, mouse liver, rat liver. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (94%), Chicken (96%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Regulates centrosome duplication, probably by inhibiting the kinase activity of ROCK2. Proposed to act as co-chaperone for HSP90. May play a role in the regulation of NOD1 via a HSP90 chaperone complex. In vitro, has intrinsic chaperone activity. This function may be achieved by inhibiting association of ROCK2 with NPM1. Involved in stress response. Prevents tumorigenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHP1
Polyclonal
Unconjugated
CHP1
1500003O03Rik; AA960066; AI046351; Cahp; calcineurin B homolog; calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin like EF-hand protein 1; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; calcium-binding protein CHP; Calcium-binding protein p22; CHP; chp1; Chromo domain-containing protein 1; chromodomain protein Chp1; EF-hand Ca2+-binding protein p22; EF-hand calcium-binding domain-containing protein p22; p22; p24; si:dkey-41n20.1; sid 470; Sid470; Sid470p; SLC9A1 binding protein; SLC9A1BP; SPAC18G6.02c; Unknown (protein for MGC:127971); Unknown (protein for MGC:63904); vac; Wrch2; wu:fc69b04; zgc:63904
Rabbit
Antigen affinity chromatography
RUO
11261, 56398, 64152
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P61022, P61023, Q99653
CHP1
Recombinant protein encompassing a sequence within the center region of human Calcium binding protein p22. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.