missing translation for 'onlineSavingsMsg'
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (rHCGa/6975) [Janelia Fluor™ 669], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-21063JF669

Product Code. 30174447

  • £508.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal antibody specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chorionic Gonadotropin beta Chain (hCG beta)
Monoclonal
Janelia Fluor 669
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB
Recombinant fragment (around aa1-116) of human Chorionic Gonadotropin beta Chain (hCG beta) protein (exact sequence is proprietary)
0.1 mL
Cancer
1082
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
rHCGa/6975
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.