missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63167UV
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.
Specifications
| hCG holo C2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CGB3 | |
| Made to Human hCG holo C2 | |
| 0.25 mL | |
| Cancer | |
| 1082 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| INN-hCG-53 | |
| Western Blot, ELISA | |
| choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction