missing translation for 'onlineSavingsMsg'
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (HCGa/2728R), DyLight 488, Novus Biologicals™

Product Code. 18375352
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375352 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375352 Supplier Bio-Techne Supplier No. NBP314273G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chorionic Gonadotropin alpha Chain (hCG alpha)
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HCGa/2728R
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-100) of human Chorionic Gonadotropin alpha Chain (hCG alpha) protein (exact sequence is proprietary) (Uniprot: P01215)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1081
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.