missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (2E6B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52442-0.025mg
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Chorionic Gonadotropin alpha Chain (HCG alpha) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
| Mouse | |
| 13 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2E6B3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CGA | |
| Purified recombinant fragment of human CGA (AA: 25-147) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 1081 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction