missing translation for 'onlineSavingsMsg'
Learn More

Chordin-like 1/CHRDL1 Antibody (263737) [DyLight 650], Novus Biologicals™

Product Code. 30733952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733952 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733952 Supplier Novus Biologicals Supplier No. FAB1808W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chordin-like 1/CHRDL1 Monoclonal antibody specifically detects Chordin-like 1/CHRDL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chordin-like 1/CHRDL1
Applications Western Blot
Classification Monoclonal
Clone 263737
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CHL, chordin-like 1, chordin-like protein 1, dA141H5.1, neuralin-1, Neurogenesin-1, NRLN1neuralin 1, ventroptin, VOPT
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chordin-like 1/CHRDL1, Glu28-Cys457, Accession # NP_001137454
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91851
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.