missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chordin Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98688B
This item is not returnable.
View return policy
Description
Chordin Polyclonal antibody specifically detects Chordin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Chordin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8646 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| CHRD, MGC133038 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Chordin | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction