missing translation for 'onlineSavingsMsg'
Learn More

Chordin Antibody (119705) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30663361 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30663361 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30663361 Supplier Novus Biologicals Supplier No. FAB7581MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Chordin Monoclonal antibody specifically detects Chordin in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chordin
Applications ELISA
Classification Monoclonal
Clone 119705
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CHRD, MGC133038
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Chordin, Thr27-Ser948, Accession # Q9Z0E2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8646
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.