missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHOP Monoclonal Antibody (K49008_7F7), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen CHOP
Clone K49008_7F7
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30202036
View Documents
Invitrogen™
MA556154
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30201314
View Documents
Invitrogen™
MA555774
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: MAAESLPFSF GTLSSWELEA WYEDLQEVLS SDENGGTYVS PPGNEEEESK IFTTLDPASL AWLTEEEPEP AEVTSTSQSP HSPDSSQSSL AQEEEEEDQG RTRKRKQSGH SPARAGKQRM KEKEQENERK VAQLAEENER LKQEIERLTR EVEATRRALI DRMVNLHQA

CHOP Monoclonal antibody specifically detects CHOP in Human samples. It is validated for Western Blot
Specifications

Specifications

CHOP
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; C/EBP-homologous protein; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; CEBPZ; CHOP; CHOP10; CHOP-10; Ddit3; DDIT-3; ddit3.L; DNA damage inducible transcript 3; DNA damage inducible transcript 3 L homeolog; DNA damage-inducible transcript 3 protein; DNA-damage inducible transcript 3; DNA-damage-inducible transcript 3; GADD153; growth arrest and DNA damage-inducible; growth arrest and DNA damage-inducible protein GADD153; Growth arrest and DNA-damage-inducible protein GADD153; MGC4154; transcription factor GADD153; Xchop; XELAEV_18013440mg
Recombinant human DDIT3 (immunogen range is 1-169).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
DDIT3
K49008_7F7
Western Blot
Unconjugated
Mouse
Human
P35638
1649
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.