missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chondrolectin Antibody (25G6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59424B
This item is not returnable.
View return policy
Description
Chondrolectin Monoclonal antibody specifically detects Chondrolectin in Human samples. It is validated for Western Blot, ELISA
Specifications
| Chondrolectin | |
| Monoclonal | |
| Biotin | |
| C21orf68, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Recombinant human Chondrolectin (22-216aa) purified from E. coli | |
| 0.1 mL | |
| Developmental Biology | |
| 140578 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 25G6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction