missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chondroitin Sulfate Monoclonal Antibody (CS-56)
GREENER_CHOICE

Product Code. 10529373
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10529373 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10529373 Supplier Invitrogen™ Supplier No. MA183055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Chondroitin sulphate antibody, clone CS-56 recognizes chondroitin sulphate types A and C (not B, dermatan sulphate).

Chondroitin sulfate is a sulfated glycosaminoglycan composed of a chain of alternating sugars. It is usually found attached to proteins as part of a proteoglycan. A chondroitin chain can have over 100 individual sugars, each of which can be sulfated in variable positions and quantities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone CS-56
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene Alias CS
Host Species Mouse
Immunogen Ventral membranes of chicken gizzard fibroblasts.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Bovine, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.