missing translation for 'onlineSavingsMsg'
Learn More

Chondroitin sulfate, MedChemExpress

Product Code. 16499885
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499885 250 mg 250mg
16452379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499885 Supplier Medchem Express Supplier No. HYB2162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chondroitin sulfate, one of five classes of glycosaminoglycans, has been widely used in the treatment of osteoarthritis. Chondroitin sulfate reduces inflammation mediators and the apoptotic process and is able to reduce protein production of inflammatory cytokines, iNOS and MMPs.

Chondroitin sulfate, one of five classes of glycosaminoglycans, has been widely used in the treatment of osteoarthritis. Chondroitin sulfate reduces inflammation mediators and the apoptotic process and is able to reduce protein production of inflammatory cytokines, iNOS and MMPs.

Specifications

Chemical Name or Material Chondroitin sulfate
Color White
CAS 9007-28-7
Physical Form Solid
Purity Grade Notes Research
Molecular Formula (C14H21NO14S)n
Synonym Chondroitin polysulfate
Solubility Information H2O : ≥ 50 mg/mL
Quantity 250 mg
SMILES O=S(O)=O.O=S(O)=O.OC([C@@H]1[C@@H](OC)[C@H](O)[C@@H](O)[C@H](O[C@@H]2[C@@H](NC(C)=O)[C@H](OC)O[C@H](CO[R])[C@@H]2O[R])O1)=O.[R].[R].[=].[=].[R2=].[R1=].[n].[or]
For Use With (Application) COVID-19-anti-virus
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.