missing translation for 'onlineSavingsMsg'
Learn More

Chondroadherin Antibody, DyLight 488, Novus Biologicals™

Product Code. 18407145 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407145 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407145 Supplier Novus Biologicals Supplier No. NBP281809G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Chondroadherin Polyclonal antibody specifically detects Chondroadherin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroadherin
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias chondroadherin, chondroadherin proteoglycan, SLRR4ACartilage leucine-rich protein
Host Species Rabbit
Immunogen Chondroadherin antibody was raised against a 17 amino acid peptide near the carboxy terminus of human Chondroadherin. The immunogen is located within the last 50 amino acids of Chondroadherin. Amino Acid Squence: ASPAKFKGQHIRDTDA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1101
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.