missing translation for 'onlineSavingsMsg'
Learn More

Choline Kinase beta Antibody, Novus Biologicals™

Product Code. 18366097 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366097 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366097 Supplier Novus Biologicals Supplier No. H00001120D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Choline Kinase beta Polyclonal antibody specifically detects Choline Kinase beta in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Kinase beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005189.2
Gene Alias CHETKcholine/ethanolamine kinase beta, CHKLcholine kinase-like, choline kinase betaEK, Choline kinase-like protein, CK, CKB, CKEKBEKBcholine/ethanolamine kinase, EC 2.7.1.32, EC 2.7.1.82, Ethanolamine kinase, Ethanolamine kinase beta
Host Species Rabbit
Immunogen CHKB (NP_005189.2, 1 a.a. - 395 a.a.) full-length human protein. MAAEATAVAGSGAVGGCLAKDGLQQSKCPDTTPKRRRASSLSRDAERRAYQWCREYLGGAWRRVQPEELRVYPVSGGLSNLLFRCSLPDHLPSVGEEPREVLLRLYGAILQGVDSLVLESVMFAILAERSLGPQLYGVFPEGRLEQYIPSRPLKTQELREPVLSAAIATKMAQFHGMEMPFTKEPHWLFGTMERYLKQIQDLPPTGLPEMNLLEMYSLKDEMGNLRKLLESTPSPVVFCHNDIQEGNILLLSEPENADSLMLVDFEYSSYNYRGFDIGNHFCEWVYDYTHEEWPFYKARPTDYPTQEQQLHFIRHYLAEAKKGETLSQEEQRKLEEDLLVEVSRYALASHFFWGLWSILQASMSTIEFGYLDYAQSRFQFYFQQKGQLTSVHSSS
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 1120
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.