missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Kinase beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85632
This item is not returnable.
View return policy
Description
Choline Kinase beta Polyclonal specifically detects Choline Kinase beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Choline Kinase beta | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CHKB | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CEWVYDYTHEEWPFYKARPTDYPTQEQQLHFIRHYLAEAKKGETLSQEEQRKLEEDLLVEVSRYALASHFFWGLWS | |
| 0.1 mL | |
| Checkpoint signaling, DNA Repair | |
| 1120 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.05mg/mL | |
| Immunohistochemistry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| CHETKcholine/ethanolamine kinase beta, CHKLcholine kinase-like, choline kinase betaEK, Choline kinase-like protein, CK, CKB, CKEKBEKBcholine/ethanolamine kinase, EC 2.7.1.32, EC 2.7.1.82, Ethanolamine kinase, Ethanolamine kinase beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction